Learn More
Colonial Medical Supply Co Inc Icare TONOLAB TV02 Tonometer, Accurate Rodent IOP Measurement, No Calibration Required, 1 each
SDP
Catalog No. NC3681832
Encompass
The Icare TONOLAB TV02 Tonometer is a handheld device that measures intraocular pressure (IOP) in rodents, ensuring quick, painless assessments without local anesthesia. It features patented rebound technology for high-precision results, requires no calibration, and utilizes single-use probes to minimize contamination risks. Ideal for glaucoma research, it streamlines workflows, enhancing safety and comfort during procedures.
Catalog No. NC3681832 Supplier Colonial Medical Supply Co Inc Supplier No. TL232
Add to Cart
Add to Cart
Request a Quote
The Icare TONOLAB TV02 Tonometer is a handheld device that measures intraocular pressure (IOP) in rodents, ensuring quick, painless assessments without local anesthesia. It features patented rebound technology for high-precision results, requires no calibration, and utilizes single-use probes to minimize contamination risks. Ideal for glaucoma research, it streamlines workflows, enhancing safety and comfort during procedures.