Learn More
Med Vet International TonoVet Pet Tonometer, J1000PET, Each
Catalog No. 5000495964
Encompass_Preferred
TONOVET Pet offers a cost-effective and user-friendly solution for monitoring intraocular pressure (IOP) in dogs only, by pet owners from the comfort of their home. Designed with reliable rebound technology for easy at-home use, this device ensures effortless and accurate measurements without the need for anesthesia or calibration. Features: • 4 measurement ranges provide a simplified way to monitor pet's IOP without worrying about normal daily fluctuations • Designed for pet owners by their veterinarian • Cost effective option for IOP home monitoring • Canine setting only • 2-year warranty
Catalog No. 50-004-95964 Supplier Med Vet International Supplier No. JOR1000PET
TONOVET Pet offers a cost-effective and user-friendly solution for monitoring intraocular pressure (IOP) in dogs only, by pet owners from the comfort of their home. Designed with reliable rebound technology for easy at-home use, this device ensures effortless and accurate measurements without the need for anesthesia or calibration. Features: • 4 measurement ranges provide a simplified way to monitor pet's IOP without worrying about normal daily fluctuations • Designed for pet owners by their veterinarian • Cost effective option for IOP home monitoring • Canine setting only • 2-year warranty