Learn More
Med Vet International TonoVet Disposable Probes, J1000D1, 100-Pack
Catalog No. 5000583908
Encompass_Preferred
TonoVet The use of tonometry to measure Intra Ocular Pressure (IOP) is the only real tool for the screening of glaucoma in veterinary patients. IOP measurements are accomplished by using instruments called tonometers which measure IOP from the outside surface of the cornea. Traditional tonometers like the Tonopen Vet measure by the “applanation” method. The instrument is repeatedly pushed against the cornea manually. The amount of force necessary to flatten the cornea is calibrated and an electronic read out is made in mm of Hg. The TonoVet utilizes a newer technology called “rebound” tonometry. The portable hand held unit is about 1/4” away from the cornea. At the push of a button, a tiny probe painlessly and rapidly reaches out and touches the cornea. Six rapid measurements are taken within seconds. Most of the time the animal will not even react and certainly no local anesthesia is required. An audible beep tells of a successful result, which is displayed digitally.
Catalog No. 50-005-83908 Supplier Med Vet International Supplier No. JOR1000D1
TonoVet The use of tonometry to measure Intra Ocular Pressure (IOP) is the only real tool for the screening of glaucoma in veterinary patients. IOP measurements are accomplished by using instruments called tonometers which measure IOP from the outside surface of the cornea. Traditional tonometers like the Tonopen Vet measure by the “applanation” method. The instrument is repeatedly pushed against the cornea manually. The amount of force necessary to flatten the cornea is calibrated and an electronic read out is made in mm of Hg. The TonoVet utilizes a newer technology called “rebound” tonometry. The portable hand held unit is about 1/4” away from the cornea. At the push of a button, a tiny probe painlessly and rapidly reaches out and touches the cornea. Six rapid measurements are taken within seconds. Most of the time the animal will not even react and certainly no local anesthesia is required. An audible beep tells of a successful result, which is displayed digitally.