Learn More
Med Vet International TonoVet Pro, J1000TPRO, Each
Catalog No. 5000495958
Encompass_Preferred
TONOVET Pro is the most versatile tonometer in iCare TONOVET's product portfolio. This flagship product is designed for high-end professionals who value premium features in accuracy and usability. Its 200 degree of positional freedom is especially useful in surgical or emergency settings. Experience faster and easier IOP measurements with the Quick Measure feature. An innovative upgrade that offers all the benefits of TONOVET's patented rebound technology but in a faster and more optimized manner. Features: • 200-degree position freedom • Quick Measure for easier and faster IOP measurements • More responsive probe • Advanced probe control for improved accuracy • Display facing the user
Catalog No. 50-004-95958 Supplier Med Vet International Supplier No. JOR1000TPRO
TONOVET Pro is the most versatile tonometer in iCare TONOVET's product portfolio. This flagship product is designed for high-end professionals who value premium features in accuracy and usability. Its 200 degree of positional freedom is especially useful in surgical or emergency settings. Experience faster and easier IOP measurements with the Quick Measure feature. An innovative upgrade that offers all the benefits of TONOVET's patented rebound technology but in a faster and more optimized manner. Features: • 200-degree position freedom • Quick Measure for easier and faster IOP measurements • More responsive probe • Advanced probe control for improved accuracy • Display facing the user