Learn More
Jorgensen Labs iCare TONOVET Pro Tonometer, Veterinary IOP Measurement, Quick Measure Function, 200-Degree Positioning Freedom
SDP
Catalog No. NC3662919
Encompass_Preferred
The iCare TONOVET Pro is an advanced veterinary tonometer designed for accurate intraocular pressure measurement in animals. It features patented rebound technology for quick, painless assessments without anesthesia, making it ideal for routine and emergency use. The device's innovative "Quick Measure" function and 200-degree positioning freedom ensure adaptability for various species, providing reliable results with a user-friendly display.
Catalog No. NC3662919 Supplier Jorgensen Labs Supplier No. J1000TPRO
Add to Cart
Add to Cart
Request a Quote
The iCare TONOVET Pro is an advanced veterinary tonometer designed for accurate intraocular pressure measurement in animals. It features patented rebound technology for quick, painless assessments without anesthesia, making it ideal for routine and emergency use. The device's innovative "Quick Measure" function and 200-degree positioning freedom ensure adaptability for various species, providing reliable results with a user-friendly display.