Learn More
StatLab Tonsil IHC Control Microscope Slides
Description
- Tonsil IHC Control Slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements
- Use these slides with confidence to evaluate stain quality and performance in histology laboratories
- StatLab control slides are created using Millennia adhesion white glass slides with 90 degree corners
- Each package includes specific staining protocol
- 25/package
- Stained slide is not included
- Tissue: Tonsil
Specifications
Specifications
| Product Type | Control Slide |
| Quantity | 25 Pk. |