Learn More
Ihc World LLC TONSIL TISSUE SLIDES, NORMAL H
SDP
Catalog No. NC0680006
Encompass_Preferred
IHC World tissue slides are prepared using top quality tissues that have been processed, embedded and sectioned following standard operating procedure. The sections were cut at 5 um and mounted on charged slides.
Catalog No. NC0680006 Supplier Ihc World LLC Supplier No. TSH5024
IHC World tissue slides are prepared using top quality tissues that have been processed, embedded and sectioned following standard operating procedure. The sections were cut at 5 um and mounted on charged slides.