Learn More
Electron Microscopy Sciences Micro-Tool Ultra-Needle, 0.25 mm, Tip 26
SDP
Catalog No. 50996653
Encompass
These tools are designed for anyone who is engaged in microscope work, miniature applications as well as specimen manipulation. Micro-Tools are the smallest known precision tools available for the laboratory and micro electronics industry. Micro-Tool consists of a threaded handle (MT1), and an interchangeable threaded base with tool tip. Tip material is hardened tool steel unless otherwise noted.
Size: 0.25 mm
Tips: Tip 26
Catalog No. 50-996-653 Supplier Electron Microscopy Sciences Supplier No. 62091260.25
These tools are designed for anyone who is engaged in microscope work, miniature applications as well as specimen manipulation. Micro-Tools are the smallest known precision tools available for the laboratory and micro electronics industry. Micro-Tool consists of a threaded handle (MT1), and an interchangeable threaded base with tool tip. Tip material is hardened tool steel unless otherwise noted.
Size: 0.25 mm
Tips: Tip 26