Learn More
Electron Microscopy Sciences Micro-Tool Micro-Chisel, 0.50 mm, Tip 3
SDP
Catalog No. 50996600
Encompass
These tools are designed for anyone who is engaged in microscope work, miniature applications as well as specimen manipulation. Micro-Tools are the smallest known precision tools available for the laboratory and micro electronics industry. Micro-Tool consists of a threaded handle (MT1), and an interchangeable threaded base with tool tip. Tip material is hardened tool steel unless otherwise noted.
Size: 0.50 mm
Tips: Tip 3
Catalog No. 50-996-600 Supplier Electron Microscopy Sciences Supplier No. 620910350
These tools are designed for anyone who is engaged in microscope work, miniature applications as well as specimen manipulation. Micro-Tools are the smallest known precision tools available for the laboratory and micro electronics industry. Micro-Tool consists of a threaded handle (MT1), and an interchangeable threaded base with tool tip. Tip material is hardened tool steel unless otherwise noted.
Size: 0.50 mm
Tips: Tip 3