Learn More
Electron Microscopy Sciences Micro-Tool Micro-Scale, 0.50 mm, Tip 7
SDP
Catalog No. 50996611
Encompass
These tools are designed for anyone who is engaged in microscope work, miniature applications as well as specimen manipulation. Micro-Tools are the smallest known precision tools available for the laboratory and micro electronics industry. Micro-Tool consists of a threaded handle (MT1), and an interchangeable threaded base with tool tip. Tip material is hardened tool steel unless otherwise noted.
Size: 0.50 mm
Tips: Tip 7
Catalog No. 50-996-611 Supplier Electron Microscopy Sciences Supplier No. 62091070.50
These tools are designed for anyone who is engaged in microscope work, miniature applications as well as specimen manipulation. Micro-Tools are the smallest known precision tools available for the laboratory and micro electronics industry. Micro-Tool consists of a threaded handle (MT1), and an interchangeable threaded base with tool tip. Tip material is hardened tool steel unless otherwise noted.
Size: 0.50 mm
Tips: Tip 7