Learn More
Electron Microscopy Sciences Carbide Micro Cutting Knife 45° with 0.5 mm tip
SDP
Catalog No. 50996689
Encompass
These unique micro cutting tools are made using the latest grinding and polishing technology producing the highest quality tool on the market today. Tools are solid carbide. Handles are interchangeable in tool handle MT1.
Tip: 0.5 mm
Catalog No. 50-996-689 Supplier Electron Microscopy Sciences Supplier No. 62094090.5
These unique micro cutting tools are made using the latest grinding and polishing technology producing the highest quality tool on the market today. Tools are solid carbide. Handles are interchangeable in tool handle MT1.
Tip: 0.5 mm