Learn More
AB Sciex LLC TOOL ASSEMBLY, ELECTRODE
Catalog No. NC1522229
Encompass
This tool is designed to facilitate removal and replacement of PA 800 Enhanced and PA800 Plus electrodes (P/N A47775).
Catalog No. NC1522229 Supplier AB Sciex LLC Supplier No. A59525
This tool is designed to facilitate removal and replacement of PA 800 Enhanced and PA800 Plus electrodes (P/N A47775).