Learn More
Grainger Reelcraft TB 03 Tool Balancer, Light Duty, Composite Housing, 1.5-3 lb Capacity, 1 ea
Catalog No. 502787526
Encompass_Preferred
The light-duty spring retractable tool balancer is designed to make tools nearly weightless, effectively reducing operator fatigue and improving overall workspace safety and ergonomics. This tool balancer features a durable composite housing and a bright zinc-plated steel cable for secure operation.
  • Increases productivity by positioning tools for immediate use with minimal operator motion.
  • Reduces potential tool damage by preventing accidental drops.
  • Includes a safety tool clip and cable stop.
  • Offers a load capacity of 1.5 lb to 3.0 lb.
  • Features permanently lubricated internal components.
  • Includes a swivel hook for mounting and a snap hook for tool attachment.
  • Supports tension adjustment via a convenient palm knob.
  • Maximum operating temperature of 150 °F (65.6 °C).
Catalog No. 50-278-7526 Supplier Grainger Supplier No. 19A879
The light-duty spring retractable tool balancer is designed to make tools nearly weightless, effectively reducing operator fatigue and improving overall workspace safety and ergonomics. This tool balancer features a durable composite housing and a bright zinc-plated steel cable for secure operation.
  • Increases productivity by positioning tools for immediate use with minimal operator motion.
  • Reduces potential tool damage by preventing accidental drops.
  • Includes a safety tool clip and cable stop.
  • Offers a load capacity of 1.5 lb to 3.0 lb.
  • Features permanently lubricated internal components.
  • Includes a swivel hook for mounting and a snap hook for tool attachment.
  • Supports tension adjustment via a convenient palm knob.
  • Maximum operating temperature of 150 °F (65.6 °C).