Learn More
MSC Kennedy Tool Drawer Liner, Polyester, Non-Slip, Black, 1 Kit
Catalog No. 5000547977
Encompass
Protect tools and reduce drawer wear with this polyester drawer liner. Designed for durability and organization, the liner features a non-slip solid surface that helps keep items securely in place within tool cases and cabinets.
  • Polyester material with PVC coating for enhanced durability
  • Non-slip solid surface prevents tool movement
  • Mold and mildew resistant for long-lasting use
  • Washable surface for easy maintenance
  • Fits a wide range of tool chest and cabinet models
Catalog No. 50-005-47977 Supplier MSC Supplier No. 55598452
Protect tools and reduce drawer wear with this polyester drawer liner. Designed for durability and organization, the liner features a non-slip solid surface that helps keep items securely in place within tool cases and cabinets.
  • Polyester material with PVC coating for enhanced durability
  • Non-slip solid surface prevents tool movement
  • Mold and mildew resistant for long-lasting use
  • Washable surface for easy maintenance
  • Fits a wide range of tool chest and cabinet models