Learn More
Grainger Huot ToolTower, Steel, HSK 100A, 54 Compartments, 3 Shelves
Catalog No. 502764937
Encompass_Preferred
The ToolTower for HSK 100A is an economical, free-standing storage unit designed to organize and protect CNC toolholders. This sturdy, three-shelf steel unit features 54 total compartments, with 18 per shelf, and utilizes heavy duty nylon inserts to prevent damage to stored toolholders. The unit is designed for floor-mounted stability and offers a modular configuration that allows for stacking additional units to expand storage capacity.
  • Provides 54 storage compartments across three shelves
  • Constructed from heavy-duty 16 gauge steel
  • Features molded nylon inserts to protect each toolholder
  • Includes a modular design that can be stacked to form a 5 or 6 shelf tower
  • Offers optional tool lock tray mounting on either side panel
  • Engineered for floor-mounted installation to ensure maximum security
Catalog No. 50-276-4937 Supplier Grainger Supplier No. 16T941
The ToolTower for HSK 100A is an economical, free-standing storage unit designed to organize and protect CNC toolholders. This sturdy, three-shelf steel unit features 54 total compartments, with 18 per shelf, and utilizes heavy duty nylon inserts to prevent damage to stored toolholders. The unit is designed for floor-mounted stability and offers a modular configuration that allows for stacking additional units to expand storage capacity.
  • Provides 54 storage compartments across three shelves
  • Constructed from heavy-duty 16 gauge steel
  • Features molded nylon inserts to protect each toolholder
  • Includes a modular design that can be stacked to form a 5 or 6 shelf tower
  • Offers optional tool lock tray mounting on either side panel
  • Engineered for floor-mounted installation to ensure maximum security