Learn More
Grainger Huot CNC SuperTower, Steel, HSK100A Taper, 78 Compartments, 1 ea
Catalog No. 502766572
Encompass_Preferred
This heavy-duty storage unit is designed to securely organize and protect HSK100A taper toolholders. Constructed for industrial environments, this tower features a durable 12 gauge steel frame and offers a 78-tool capacity across three shelves.
  • Constructed of 12 gauge steel for heavy duty use
  • Finished in a baked-on gold vein powder coating for durability
  • Includes phosphate pretreatment undercoating for added toughness
  • Features angled shelving for improved tool access
  • Stackable two high to expand storage capacity
  • Dimensions: 33.75 in. W x 17.5 in. D x 33.5 in. H
  • Made in the United States
Catalog No. 50-276-6572 Supplier Grainger Supplier No. 16T920
This heavy-duty storage unit is designed to securely organize and protect HSK100A taper toolholders. Constructed for industrial environments, this tower features a durable 12 gauge steel frame and offers a 78-tool capacity across three shelves.
  • Constructed of 12 gauge steel for heavy duty use
  • Finished in a baked-on gold vein powder coating for durability
  • Includes phosphate pretreatment undercoating for added toughness
  • Features angled shelving for improved tool access
  • Stackable two high to expand storage capacity
  • Dimensions: 33.75 in. W x 17.5 in. D x 33.5 in. H
  • Made in the United States