Learn More
Grainger Tool Holder 3 Shelf Steel HSK 63A 135 pc
Catalog No. 502765559
Encompass_Preferred
The Grainger 16T921 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is thermally protected and rated for continuous duty applications. It features a cast iron frame, end brackets and conduit box. The motor is CSA certified and UL listed.
Catalog No. 50-276-5559 Supplier Grainger Supplier No. 16T921
The Grainger 16T921 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is thermally protected and rated for continuous duty applications. It features a cast iron frame, end brackets and conduit box. The motor is CSA certified and UL listed.