Learn More
Grainger Huot CNC Tool Platform, 5C, 48-Piece Capacity, Steel
Catalog No. 502766040
Encompass_Preferred
This angled CNC tool platform is designed to store 48 5C collets, providing organized and accessible storage for machining environments. Its angled design ensures clear visibility and efficient selection of tools, making it an ideal solution for benchtop organization.
  • Holds 48 5C collets
  • Constructed from heavy gauge steel for durability
  • Features a tough epoxy finish for long-lasting protection
  • Includes heavy-duty nylon inserts to prevent damage to tooling
  • Designed for benchtop mounting for improved access and efficiency
Catalog No. 50-276-6040 Supplier Grainger Supplier No. 16T917
This angled CNC tool platform is designed to store 48 5C collets, providing organized and accessible storage for machining environments. Its angled design ensures clear visibility and efficient selection of tools, making it an ideal solution for benchtop organization.
  • Holds 48 5C collets
  • Constructed from heavy gauge steel for durability
  • Features a tough epoxy finish for long-lasting protection
  • Includes heavy-duty nylon inserts to prevent damage to tooling
  • Designed for benchtop mounting for improved access and efficiency