Learn More
Grainger Huot Machine Mount Rack, 40 Taper, Steel, 28 Tools, 1 ea
Catalog No. 502784906
Encompass_Preferred
This compact machine mount tool rack provides organized, high-capacity storage for 40 taper toolholders. Designed for efficiency, the rack features a durable steel construction with a scratch-resistant powder coat finish. Its low-profile design protrudes only 11 in. from the machine or wall, and it offers versatile configuration options to optimize tool accessibility and workspace organization.
  • Holds up to 28 tools
  • Includes nylon inserts to protect and securely hold toolholders
  • Features 14 slots per side for adjustable shelf height
  • Shelves mount flat or at a -20° angle for ergonomic tool presentation
  • Constructed with a 14 gauge steel base and 16 gauge steel shelves
  • Equipped with 6 mounting locations for a sturdy attachment
Catalog No. 50-278-4906 Supplier Grainger Supplier No. 18EC65
This compact machine mount tool rack provides organized, high-capacity storage for 40 taper toolholders. Designed for efficiency, the rack features a durable steel construction with a scratch-resistant powder coat finish. Its low-profile design protrudes only 11 in. from the machine or wall, and it offers versatile configuration options to optimize tool accessibility and workspace organization.
  • Holds up to 28 tools
  • Includes nylon inserts to protect and securely hold toolholders
  • Features 14 slots per side for adjustable shelf height
  • Shelves mount flat or at a -20° angle for ergonomic tool presentation
  • Constructed with a 14 gauge steel base and 16 gauge steel shelves
  • Equipped with 6 mounting locations for a sturdy attachment