Learn More
Grainger Huot CNC Tool Rack, HSK63A, Steel, 28 Capacity, 1 ea
Catalog No. 502784631
Encompass_Preferred
This compact machine mount tool rack is designed for efficient organization of HSK63A taper toolholders directly on the machine. Constructed from heavy-duty 14-gauge steel with 16-gauge steel shelves and a durable, scratch-resistant powder-coated finish, it provides a stable and long-lasting storage solution.
  • Holds up to 28 HSK63A, HSK80B, and HSK80D toolholders
  • Includes protective nylon inserts to securely hold toolholders
  • Features 14 slots per side for flexible shelf height adjustments
  • Shelves mount flat or at a -20° angle for optimal tool presentation
  • Utilizes 6 mounting locations for a sturdy attachment to the machine
  • Low profile design protrudes only 11 in. from the machine
Catalog No. 50-278-4631 Supplier Grainger Supplier No. 18EC67
This compact machine mount tool rack is designed for efficient organization of HSK63A taper toolholders directly on the machine. Constructed from heavy-duty 14-gauge steel with 16-gauge steel shelves and a durable, scratch-resistant powder-coated finish, it provides a stable and long-lasting storage solution.
  • Holds up to 28 HSK63A, HSK80B, and HSK80D toolholders
  • Includes protective nylon inserts to securely hold toolholders
  • Features 14 slots per side for flexible shelf height adjustments
  • Shelves mount flat or at a -20° angle for optimal tool presentation
  • Utilizes 6 mounting locations for a sturdy attachment to the machine
  • Low profile design protrudes only 11 in. from the machine