Learn More
Grainger Tool Holder Steel C4 CAPTO CNC Tools
Catalog No. 502765052
Encompass_Preferred
The Grainger 16T943 is a 3M Scotch-Brite General Purpose Hand Pad with an open web construction and non-woven fibers in medium grade for cleaning and scouring. This 6x9 inch pad is ideal for cleaning pots, pans, dishes, sinks, showers, tubs, stoves, ovens, grills, concrete, and metal. It removes grease, grime, rust stains, and soap scum without scratching. The durable fibers resist shredding, and finger straps on the back provide an abrasive surface for tough cleaning jobs.
Catalog No. 50-276-5052 Supplier Grainger Supplier No. 16T943
The Grainger 16T943 is a 3M Scotch-Brite General Purpose Hand Pad with an open web construction and non-woven fibers in medium grade for cleaning and scouring. This 6x9 inch pad is ideal for cleaning pots, pans, dishes, sinks, showers, tubs, stoves, ovens, grills, concrete, and metal. It removes grease, grime, rust stains, and soap scum without scratching. The durable fibers resist shredding, and finger straps on the back provide an abrasive surface for tough cleaning jobs.