Learn More
Grainger Huot CNC ToolTower, Steel, 3 Shelves, 72-Piece Capacity, 1 ea
Catalog No. 502765348
Encompass_Preferred
The ToolTower for Coromant CAPTO C5 tooling is an economical free-standing storage unit designed to store and protect CNC toolholders in a compact footprint. This industrial tower is constructed from sturdy 16 gauge steel and features a durable epoxy finish for long-lasting use in demanding environments.
  • Designed for Coromant CAPTO C5 tooling
  • Holds up to 72 pieces
  • Features 3 shelves for organized storage
  • Constructed from durable 16 gauge steel
  • Dimensions measure 27.3125 in. wide by 18.3125 in. deep by 36 in. high
  • Security can be enhanced by bolting the unit to the floor
Catalog No. 50-276-5348 Supplier Grainger Supplier No. 16T944
The ToolTower for Coromant CAPTO C5 tooling is an economical free-standing storage unit designed to store and protect CNC toolholders in a compact footprint. This industrial tower is constructed from sturdy 16 gauge steel and features a durable epoxy finish for long-lasting use in demanding environments.
  • Designed for Coromant CAPTO C5 tooling
  • Holds up to 72 pieces
  • Features 3 shelves for organized storage
  • Constructed from durable 16 gauge steel
  • Dimensions measure 27.3125 in. wide by 18.3125 in. deep by 36 in. high
  • Security can be enhanced by bolting the unit to the floor