Learn More
Grainger Huot CNC ToolTower, Steel, 72 Compartments, CAPTO C6, 1 ea
Catalog No. 502768334
Encompass_Preferred
The CNC ToolTower provides organized storage for CAPTO (C6) taper tools. Constructed from durable 16 gauge steel, this tower features 3 shelves with 72 total compartments, allowing for a capacity of 24 tools per shelf. The unit comes in a red finish and includes a tool pouch for added utility.
  • Material: Steel
  • Capacity: 72 tools (24 per shelf)
  • Compatibility: CAPTO (C6) taper tools
  • Dimensions: 27-5/16 in. (width) x 36 in. (height) x 18-5/16 in. (depth)
  • Construction: Sturdy 16 gauge steel
  • Additional features: Optional tool lock tray can be mounted on either side panel; unit should be bolted to the floor for additional security
Catalog No. 50-276-8334 Supplier Grainger Supplier No. 16T945
The CNC ToolTower provides organized storage for CAPTO (C6) taper tools. Constructed from durable 16 gauge steel, this tower features 3 shelves with 72 total compartments, allowing for a capacity of 24 tools per shelf. The unit comes in a red finish and includes a tool pouch for added utility.
  • Material: Steel
  • Capacity: 72 tools (24 per shelf)
  • Compatibility: CAPTO (C6) taper tools
  • Dimensions: 27-5/16 in. (width) x 36 in. (height) x 18-5/16 in. (depth)
  • Construction: Sturdy 16 gauge steel
  • Additional features: Optional tool lock tray can be mounted on either side panel; unit should be bolted to the floor for additional security