Learn More
Grainger Huot Tool Holder Cart, Steel, HSK100A Taper, 36 Compartments, 1 ea.
Catalog No. 502787117
Encompass_Preferred
This locking steel cart is designed for the secure storage and organization of HSK100A taper toolholders. Featuring 36 compartments with molded inserts, the unit prevents tool wear and ensures accessibility while keyed doors provide protection against unauthorized access. Transparent doors allow for content visibility without needing to unlock the unit.
  • Holds up to 36 HSK100A taper toolholders
  • Features durable steel construction with a powder coated finish
  • Includes heavy duty 5 in. locking casters for mobility
  • Equipped with two sets of doors to secure both sides of the cart
  • Includes a large open top tray area for utility transport
  • Includes four adjustable shelves with seven mounting positions
  • Includes a side tray for optional tool changing vises or collet racks
  • Offers a weight capacity of 1,000 lb.
Catalog No. 50-278-7117 Supplier Grainger Supplier No. 18EC72
This locking steel cart is designed for the secure storage and organization of HSK100A taper toolholders. Featuring 36 compartments with molded inserts, the unit prevents tool wear and ensures accessibility while keyed doors provide protection against unauthorized access. Transparent doors allow for content visibility without needing to unlock the unit.
  • Holds up to 36 HSK100A taper toolholders
  • Features durable steel construction with a powder coated finish
  • Includes heavy duty 5 in. locking casters for mobility
  • Equipped with two sets of doors to secure both sides of the cart
  • Includes a large open top tray area for utility transport
  • Includes four adjustable shelves with seven mounting positions
  • Includes a side tray for optional tool changing vises or collet racks
  • Offers a weight capacity of 1,000 lb.