Learn More
Grainger Huot SecurityScoot, 50 Taper, Steel, 36 Compartments, 1 ea
Catalog No. 502784717
Encompass_Preferred
The SecurityScoot 50 Taper CNC tool cart provides durable storage and mobility for shop environments. Constructed from steel with a scratch-resistant powder coat finish, this cart secures and organizes up to 36 toolholders. It features clear polycarbonate panels on the doors for immediate tool identification and keyed-alike locks to ensure safety in demanding shop conditions.
  • Holds 36 BT50, CAT50, or NMTB50 taper toolholders
  • Adjustable shelves with 7 mounting positions
  • Lockable doors with clear panels for visibility
  • Heavy duty 5 in. locking casters for stability
  • Side tray included for optional vise or collet rack mounting
  • Large open top tray for utility transport
  • Sturdy steel construction with powder coated finish
Catalog No. 50-278-4717 Supplier Grainger Supplier No. 18EC70
The SecurityScoot 50 Taper CNC tool cart provides durable storage and mobility for shop environments. Constructed from steel with a scratch-resistant powder coat finish, this cart secures and organizes up to 36 toolholders. It features clear polycarbonate panels on the doors for immediate tool identification and keyed-alike locks to ensure safety in demanding shop conditions.
  • Holds 36 BT50, CAT50, or NMTB50 taper toolholders
  • Adjustable shelves with 7 mounting positions
  • Lockable doors with clear panels for visibility
  • Heavy duty 5 in. locking casters for stability
  • Side tray included for optional vise or collet rack mounting
  • Large open top tray for utility transport
  • Sturdy steel construction with powder coated finish