Learn More
Grainger Huot CNC Wall Tree Locker, Steel, 30 Toolholder Capacity, 1 ea
Catalog No. 502786911
Encompass_Preferred
The Tool Holder Wall Locker is a rugged steel storage cabinet designed to secure, organize, and protect 50 taper toolholders from dust, debris, and theft. The unit features reinforced locking doors and a durable powder-coated finish suitable for production shop environments.
  • Holds 30 toolholders
  • Constructed from 12 and 16 gauge steel
  • Includes three shelves that can be mounted flat or at a -20° angle
  • Features nylon inserts to protect and securely hold toolholders
  • Shelves are interchangeable with other company storage systems
  • Compatible with BT50 and CAT50 taper tool holders
Catalog No. 50-278-6911 Supplier Grainger Supplier No. 18EC74
The Tool Holder Wall Locker is a rugged steel storage cabinet designed to secure, organize, and protect 50 taper toolholders from dust, debris, and theft. The unit features reinforced locking doors and a durable powder-coated finish suitable for production shop environments.
  • Holds 30 toolholders
  • Constructed from 12 and 16 gauge steel
  • Includes three shelves that can be mounted flat or at a -20° angle
  • Features nylon inserts to protect and securely hold toolholders
  • Shelves are interchangeable with other company storage systems
  • Compatible with BT50 and CAT50 taper tool holders