Learn More
Grainger Huot Workstation, 50 Taper, Steel, 18-Tool Capacity, 1 ea
Catalog No. 502786913
Encompass_Preferred
The workstation provides a durable, organized storage solution for machine shop environments, featuring a hardwood butcher block top and heavy-duty welded steel construction. Designed for efficiency, it keeps tools and accessories accessible while offering secure, lockable storage to protect against debris.
  • Durable hardwood butcher block top with ample workspace
  • Full extension ball bearing drawer slides
  • Locking doors to secure tools and protect against chips and debris
  • Two top drawers with welded columns and 1 in. dividers for customizable storage
  • Larger 4 in. bottom drawer for miscellaneous tools
  • Bottom tooling drawer capable of holding up to 30 toolholders
  • Threaded side holes for mounting a tool changing vise horizontally or vertically
  • Integrated forklift pockets for mobility
Catalog No. 50-278-6913 Supplier Grainger Supplier No. 18EC62
The workstation provides a durable, organized storage solution for machine shop environments, featuring a hardwood butcher block top and heavy-duty welded steel construction. Designed for efficiency, it keeps tools and accessories accessible while offering secure, lockable storage to protect against debris.
  • Durable hardwood butcher block top with ample workspace
  • Full extension ball bearing drawer slides
  • Locking doors to secure tools and protect against chips and debris
  • Two top drawers with welded columns and 1 in. dividers for customizable storage
  • Larger 4 in. bottom drawer for miscellaneous tools
  • Bottom tooling drawer capable of holding up to 30 toolholders
  • Threaded side holes for mounting a tool changing vise horizontally or vertically
  • Integrated forklift pockets for mobility