Learn More
Grainger Huot Tool Holder Workstation, 40 Taper, Steel, 30-Tool Capacity
Catalog No. 502787084
Encompass_Preferred
This workstation is designed for efficient organization and storage of 40 taper toolholders in machine shop environments. Constructed from cold rolled steel with a durable wood work surface, this unit provides a stable, accessible station for operators to manage tools and daily tasks. It features secure locking storage and is designed to integrate into high-traffic workshop areas.
  • Holds 30 toolholders
  • Compatible with 40 taper tool holders including BT40, CAT40, and NMTB40
  • Includes two drawers with full extension ball bearing slides
  • Features cabinet pockets for forklift movement
  • Includes locking doors to protect tools from chips and debris
  • Pre-drilled on both sides for horizontal or vertical tool changing vise mounting
  • Large wood top surface for daily tasks
Catalog No. 50-278-7084 Supplier Grainger Supplier No. 18EC61
This workstation is designed for efficient organization and storage of 40 taper toolholders in machine shop environments. Constructed from cold rolled steel with a durable wood work surface, this unit provides a stable, accessible station for operators to manage tools and daily tasks. It features secure locking storage and is designed to integrate into high-traffic workshop areas.
  • Holds 30 toolholders
  • Compatible with 40 taper tool holders including BT40, CAT40, and NMTB40
  • Includes two drawers with full extension ball bearing slides
  • Features cabinet pockets for forklift movement
  • Includes locking doors to protect tools from chips and debris
  • Pre-drilled on both sides for horizontal or vertical tool changing vise mounting
  • Large wood top surface for daily tasks