Learn More
MSC Bosch Power Tool Hose Adapter, Composite, Quick Click System, Air Regulating Slider
Catalog No. 502835667
Encompass
The Power Tool Hose Adapter is designed for efficient dust extraction, connecting to various tools and attachments. It features a rubber-nozzle tool end for a secure friction fit and an air regulating slider to control airflow, which is particularly beneficial for applications like sanding where reduced airflow prevents the tool from sticking to the workpiece. Its quick click feature ensures a secure attachment to the hose, and the nozzle is designed for a comfortable grip.
  • Connects to hose with the quick click system.
  • Rubber nozzle sized for a friction fit on most dust collection attachments.
  • Air regulating slider controls the amount of air flow.
  • Prevents tool from sticking to the workpiece during sanding by reducing airflow.
  • Quick click feature for secure attachment.
  • Nozzle designed for a comfortable grip.
Catalog No. 50-283-5667 Supplier MSC Supplier No. 47842166
The Power Tool Hose Adapter is designed for efficient dust extraction, connecting to various tools and attachments. It features a rubber-nozzle tool end for a secure friction fit and an air regulating slider to control airflow, which is particularly beneficial for applications like sanding where reduced airflow prevents the tool from sticking to the workpiece. Its quick click feature ensures a secure attachment to the hose, and the nozzle is designed for a comfortable grip.
  • Connects to hose with the quick click system.
  • Rubber nozzle sized for a friction fit on most dust collection attachments.
  • Air regulating slider controls the amount of air flow.
  • Prevents tool from sticking to the workpiece during sanding by reducing airflow.
  • Quick click feature for secure attachment.
  • Nozzle designed for a comfortable grip.