Learn More
Fine Science Tools Guthrie Retractor, Stainless Steel, 13 cm
SDP
Catalog No. NC0074364
Encompass
Product: Guthrie Retractor - 2 Pronged Blunt/13cm/ Teeth: 2 Pronged Blunt/ Teeth Width: 2.5mm/ Teeth Depth: 3mm/ Alloy: Stainless Steel/ Length: 13cm/ Unit: Each
Catalog No. NC0074364 Supplier Fine Science Tools Supplier No. 1702113
Product: Guthrie Retractor - 2 Pronged Blunt/13cm/ Teeth: 2 Pronged Blunt/ Teeth Width: 2.5mm/ Teeth Depth: 3mm/ Alloy: Stainless Steel/ Length: 13cm/ Unit: Each