Learn More
Grainger Tool Safety Cover 14x5x14 In Clear
Catalog No. 502768282
Encompass_Preferred
The HUOT Tool Safety Cover is a durable plastic cover designed to protect tools and equipment. It measures 12 inches wide x 18 inches long x 6 inches high and features a hinged lid for easy access. The cover is made from high-density polyethylene plastic to withstand impacts and has ventilation holes to allow airflow. It can accommodate most hand tools up to 12 inches long. The HUOT safety cover comes in orange for high visibility and has a latch to securely close the lid. This tool box helps organize, transport, and protect shop tools and power tool accessories.
Catalog No. 50-276-8282 Supplier Grainger Supplier No. 16T936
The HUOT Tool Safety Cover is a durable plastic cover designed to protect tools and equipment. It measures 12 inches wide x 18 inches long x 6 inches high and features a hinged lid for easy access. The cover is made from high-density polyethylene plastic to withstand impacts and has ventilation holes to allow airflow. It can accommodate most hand tools up to 12 inches long. The HUOT safety cover comes in orange for high visibility and has a latch to securely close the lid. This tool box helps organize, transport, and protect shop tools and power tool accessories.