Learn More
Grainger Tool Suspension Frame 60Wx18Dx6 H Beige
Catalog No. 502768100
Encompass_Preferred
The Grainger 16Y945 is a 3M Scotch-Brite General Purpose Hand Pad that features an open web construction and is made of nylon and synthetic fibers in a non-woven material. This 8" x 4" hand pad has a medium duty scrubbing power and is ideal for cleaning pots, pans, dishes, flatware, and other kitchenware. It effectively removes dried or burnt on foods. The open web design rinses easily to minimize cross contamination. It can be used wet or dry. This hand pad is latex-free, disposable, and intended for medium duty scrubbing.
Catalog No. 50-276-8100 Supplier Grainger Supplier No. 16Y945
The Grainger 16Y945 is a 3M Scotch-Brite General Purpose Hand Pad that features an open web construction and is made of nylon and synthetic fibers in a non-woven material. This 8" x 4" hand pad has a medium duty scrubbing power and is ideal for cleaning pots, pans, dishes, flatware, and other kitchenware. It effectively removes dried or burnt on foods. The open web design rinses easily to minimize cross contamination. It can be used wet or dry. This hand pad is latex-free, disposable, and intended for medium duty scrubbing.