Learn More
Grainger Tool Tote Rack Steel 50 Taper Tools 5 Pc
Catalog No. 502768335
Encompass_Preferred
The HUOT Tool Tote Rack is a benchtop-mounted storage system for organizing hand tools and small parts. It is constructed of steel with a durable powder coat finish and has 18 removable plastic bins. The bins have ventilation slots to allow air circulation and are clear so contents are visible. The rack measures 25"W x 6"D x 12"H and can hold up to 50 lbs. It mounts to benches up to 1.25" thick using the included hardware. This made-in-the-USA rack keeps frequently used tools and supplies within reach to save time and improve workflow.
Catalog No. 50-276-8335 Supplier Grainger Supplier No. 16T948
Add to Cart
Add to Cart
The HUOT Tool Tote Rack is a benchtop-mounted storage system for organizing hand tools and small parts. It is constructed of steel with a durable powder coat finish and has 18 removable plastic bins. The bins have ventilation slots to allow air circulation and are clear so contents are visible. The rack measures 25"W x 6"D x 12"H and can hold up to 50 lbs. It mounts to benches up to 1.25" thick using the included hardware. This made-in-the-USA rack keeps frequently used tools and supplies within reach to save time and improve workflow.