Learn More
MSC Sellars Wipers, WATERWEAVE T600, Jumbo Roll, White, 1,100 Sheets
Catalog No. 502838046
Encompass
These WaterWeave T600 wipers are engineered for medium-duty, wet or dry wiping tasks, effectively handling tough messes, solvent application, and hand degreasing. Utilizing WaterWeave technology, polypropylene and cellulose fibers are hydroentangled for enhanced strength, durability, and extended usage, resulting in smooth, highly absorbent, and low-lint performance.
  • Designed for medium-duty, wet or dry wiping tasks.
  • Ideal for applications using solvents, cleaning rough surfaces, or requiring low lint.
  • Jumbo rolls fit wall mount or floor stand dispensers.
  • Features superior strength and durability from woven fibers.
  • Low-linting, latex-free, and contaminant-free.
  • Suitable for equipment cleaning, surface preparation, and maintenance.
Catalog No. 50-283-8046 Supplier MSC Supplier No. 80595929
These WaterWeave T600 wipers are engineered for medium-duty, wet or dry wiping tasks, effectively handling tough messes, solvent application, and hand degreasing. Utilizing WaterWeave technology, polypropylene and cellulose fibers are hydroentangled for enhanced strength, durability, and extended usage, resulting in smooth, highly absorbent, and low-lint performance.
  • Designed for medium-duty, wet or dry wiping tasks.
  • Ideal for applications using solvents, cleaning rough surfaces, or requiring low lint.
  • Jumbo rolls fit wall mount or floor stand dispensers.
  • Features superior strength and durability from woven fibers.
  • Low-linting, latex-free, and contaminant-free.
  • Suitable for equipment cleaning, surface preparation, and maintenance.