Learn More
Electron Microscopy Sciences Tooled Neck Vials, 1 Dram
SDP
Catalog No. 501899978
Encompass_Preferred
These vials are made from Type 1, Class B borosilicate glass. Their tooled necks ensure accurate fit with "snap-in" closures. They are ideal for storage of specimens and sample blocks. Comes with an open-bottom closure.
Volume: 1 Dram
Catalog No. 50-189-9978 Supplier Electron Microscopy Sciences Supplier No. 7264601
These vials are made from Type 1, Class B borosilicate glass. Their tooled necks ensure accurate fit with "snap-in" closures. They are ideal for storage of specimens and sample blocks. Comes with an open-bottom closure.
Volume: 1 Dram