Learn More
Electron Microscopy Sciences Tooled Neck Vials, 5 Dram
SDP
Catalog No. 501899980
Encompass_Preferred
These vials are made from Type 1, Class B borosilicate glass. Their tooled necks ensure accurate fit with "snap-in" closures. They are ideal for storage of specimens and sample blocks. Comes with a closed bottom, two-piece closure.
Volume: 5 Dram
Catalog No. 50-189-9980 Supplier Electron Microscopy Sciences Supplier No. 7264605
These vials are made from Type 1, Class B borosilicate glass. Their tooled necks ensure accurate fit with "snap-in" closures. They are ideal for storage of specimens and sample blocks. Comes with a closed bottom, two-piece closure.
Volume: 5 Dram