Learn More
Electron Microscopy Sciences Tooled Neck Vials, 7 Dram
SDP
Catalog No. 50949572
Encompass_Preferred
These vials are made from Type 1, Class B borosilicate glass. Their tooled necks ensure accurate fit with "snap-in" closures. They are ideal for storage of specimens and sample blocks. Comes with a closed bottom, two-piece closure.
Volume: 7 Dram
Catalog No. 50-949-572 Supplier Electron Microscopy Sciences Supplier No. 7264607
These vials are made from Type 1, Class B borosilicate glass. Their tooled necks ensure accurate fit with "snap-in" closures. They are ideal for storage of specimens and sample blocks. Comes with a closed bottom, two-piece closure.
Volume: 7 Dram