Learn More
Grainger Toolholder Vise Steel BT 40 Taper Tools
Catalog No. 502767105
Encompass_Preferred
The HUOT 200T41 CNC Toolholder Vise is a precision toolholding vise designed for CNC machining applications. It features a hardened steel body with a 200mm x 41mm jaw opening capacity. The vise has a rigid one-piece body construction and comes with removable and reversible steel jaws. It can hold tools up to 1.5 inches in diameter. The vise has a low profile design to provide increased Z axis clearance. It comes with mounting holes for easy installation on a CNC machine table. The vise weighs 11 lbs and has an overall height of 3.5 inches.
Catalog No. 50-276-7105 Supplier Grainger Supplier No. 16T925
The HUOT 200T41 CNC Toolholder Vise is a precision toolholding vise designed for CNC machining applications. It features a hardened steel body with a 200mm x 41mm jaw opening capacity. The vise has a rigid one-piece body construction and comes with removable and reversible steel jaws. It can hold tools up to 1.5 inches in diameter. The vise has a low profile design to provide increased Z axis clearance. It comes with mounting holes for easy installation on a CNC machine table. The vise weighs 11 lbs and has an overall height of 3.5 inches.