Learn More
Grainger Toolholder Vise Stl CAT-V 40 Taper Tools
Catalog No. 502766151
Encompass_Preferred
The HUOT Toolholder Vise for CAT40 is a toolholder vise designed for CAT40 spindles. It has a clamping range of 0.5" to 1.5" and can hold tool shanks up to 1.5" in diameter. The vise body is made from ductile iron and has an opening stroke of 2.25". It comes with removable steel jaws and uses a screw clamping mechanism for rigid clamping. The vise can withstand speeds up to 4000 RPM. Overall dimensions are 4" x 3.25" x 4.5". It weighs approximately 9 lbs.
Catalog No. 50-276-6151 Supplier Grainger Supplier No. 16T924
The HUOT Toolholder Vise for CAT40 is a toolholder vise designed for CAT40 spindles. It has a clamping range of 0.5" to 1.5" and can hold tool shanks up to 1.5" in diameter. The vise body is made from ductile iron and has an opening stroke of 2.25". It comes with removable steel jaws and uses a screw clamping mechanism for rigid clamping. The vise can withstand speeds up to 4000 RPM. Overall dimensions are 4" x 3.25" x 4.5". It weighs approximately 9 lbs.