Learn More
Cayman Chemical Toosendanin 10mg
SDP
Catalog No. 502949573
Encompass_Preferred
A triterpenoid with diverse biological activities; inhibits proliferation of BEL-7404, U251, SH-SY5Y, HL-60, PC3, and U937 cancer cells (IC50s = A. aegypti first instar larvae (LC50 = 60.8 UG/ml) and delays pupal development at 6.3-25 UG/ml; decreases serum levels of TNF-α, IL-1β, and IL-6 in a mouse model of ulcerative colitis induced by DSS at 0.5 and 1 mg/kg; increases survival in a mouse model of BoNT/A-induced botulism at 9 mg/kg
Catalog No. 50-294-9573 Supplier Cayman Chemical Supplier No. 3404210 MG
A triterpenoid with diverse biological activities; inhibits proliferation of BEL-7404, U251, SH-SY5Y, HL-60, PC3, and U937 cancer cells (IC50s = A. aegypti first instar larvae (LC50 = 60.8 UG/ml) and delays pupal development at 6.3-25 UG/ml; decreases serum levels of TNF-α, IL-1β, and IL-6 in a mouse model of ulcerative colitis induced by DSS at 0.5 and 1 mg/kg; increases survival in a mouse model of BoNT/A-induced botulism at 9 mg/kg