Learn More
Surgical Design™ Tooth Extracting Forceps - Multiple Patterns Available
Catalog No. 21124743 Shop All Surgical Design Products
Change view
Click to view available options
Chuck Size:
No.150
No.17
No.210
No.222
No.53L
No.53R
No.74
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chuck Size
21-124-743 No.150
21-124-744 No.17
21-124-745 No.210
21-124-746 No.222
21-124-747 No.53L
21-124-748 No.53R
21-124-749 No.74
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 21-124-743 Supplier Surgical Design™ Supplier No. EXF150
Only null left
Add to Cart
Add to Cart
Tooth extracting forceps are engineered for firm, controlled grasping during extractions across dental and surgical applications.
  • Available Patterns: 150 Universal upper forceps, 17 Lower molars, 210 Upper molars, 222 Lower third molars, 53L Upper molars left, 53R Upper molars right, 74 Lower incisors and roots
  • Designed for firm grasping and controlled extraction of teeth and roots
  • Anatomically shaped beaks tailored to specific tooth locations
  • Strong hinge and precision alignment ensure stable, slip-free operation
  • Textured, ergonomic handles minimize hand fatigue and improve leverage
  • Manufactured from high-grade surgical stainless steel for durability
  • Fully autoclavable and corrosion-resistant
  • Ideal for general dentistry, oral surgery, emergency extraction, and forensic dentistry
  • Autopsy/postmortem applications: Removal of teeth for identification, jaw access, root and fragment extraction during forensic examinations
Specifications
Product Type Extraction Forceps
Chuck Size No.150
Handle Material Stainless Steel
Length (Metric) 17.8 cm
Material Stainless Steel
Size Adult
Autoclavable Autoclavable
Color Silver