Learn More
Med Vet International Tooth Punch-Straight, 20cm, J0712
Catalog No. 5000584582
Encompass_Preferred
The punch is used for removal of cheek teeth through a trephined hole into the sinus. A knurled handle gives a better grip. A round head with a slight recessed area fits better onto the tooth. Stainless steel. 20cm L.
Catalog No. 50-005-84582 Supplier Med Vet International Supplier No. JOR712
The punch is used for removal of cheek teeth through a trephined hole into the sinus. A knurled handle gives a better grip. A round head with a slight recessed area fits better onto the tooth. Stainless steel. 20cm L.