Learn More
Med Vet International Toothbrush Colgate Adult Soft, 11905676, Each
Catalog No. 503044115
Encompass_Preferred
Adult Soft bristle with pink straight handle Reusable oral hygiene instrument for patients with difficulty swallowing and expectorating, or who cannot perform routine oral care Remove mucus and food build-up, as well as plaque, bacteria and fluid Help eliminate chronic halitosis Reduce the risk of infection, aspiration and pneumonia Ideally suited for use with both wall suction and portable pumps
Catalog No. 50-304-4115 Supplier Med Vet International Supplier No. 1123098EA
Adult Soft bristle with pink straight handle Reusable oral hygiene instrument for patients with difficulty swallowing and expectorating, or who cannot perform routine oral care Remove mucus and food build-up, as well as plaque, bacteria and fluid Help eliminate chronic halitosis Reduce the risk of infection, aspiration and pneumonia Ideally suited for use with both wall suction and portable pumps