Learn More
Fine Science Tools Extra Fine Graefe Forceps, Stainless Steel, 10 cm
SDP
Catalog No. NC9746059
Encompass
Tip Shape: Straight/ Tips: 1x2 Teeth/ Tip Dimensions: 0.5 x 0.5mm/ Alloy / Material: Stainless Steel/ Length: 10cm
Catalog No. NC9746059 Supplier Fine Science Tools Supplier No. 1115310
Tip Shape: Straight/ Tips: 1x2 Teeth/ Tip Dimensions: 0.5 x 0.5mm/ Alloy / Material: Stainless Steel/ Length: 10cm