Learn More
MOLTOX Molecular Toxicology Inc Top Agar, 0.6% Agar & 5% NaCl, 500 mL/bottle, Store at Room Temperature
SDP
Catalog No. 50948787
Encompass_Preferred
For use with Minimal Glucose Agar, S. typhimurium and E. coli WP2 strains in the Ames assay. Requires supplementation. MOLTOX prepared media and media components are specifically formulated for use in bacterial mutagenicity assays. Minimal Glucose agars, top agars, master plates, nutrient broth and phenotype confirmation media are prepared with various formulations for specialized laboratory needs.
Catalog No. 50-948-787 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 26632.5
For use with Minimal Glucose Agar, S. typhimurium and E. coli WP2 strains in the Ames assay. Requires supplementation. MOLTOX prepared media and media components are specifically formulated for use in bacterial mutagenicity assays. Minimal Glucose agars, top agars, master plates, nutrient broth and phenotype confirmation media are prepared with various formulations for specialized laboratory needs.