Learn More
BINDER™ Top Access Port (10 mm)
SDP
Catalog No. 08111488 Shop All BINDER Inc. Products
Change view
Click to view available options
Diameter (Metric) Inside:
10 mm
Catalog No. Diameter (Metric) Inside
08-111-488 10 mm
1 options
Catalog No. 08-111-488 Supplier BINDER™ Supplier No. 80121261
Add to Cart
Add to Cart
For Use With: BF 56, BF 115, BF 260, BF 400, BF 720
  • For use on top
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
Description Access port with silicone plug - 10 mm, top
For Use With (Equipment) BINDER Chambers BF 56, BF 115, BF 260, BF 400, BF 720
Material Steel
Type Access Port
Includes Silicone plug
Diameter (Metric) Inside 10 mm