Learn More
BINDER™ Top Access Port (10 mm)
SDP
Catalog No. 08111491 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
BINDER Chambers Series FED
Catalog No. For Use With (Equipment)
08-111-491 BINDER Chambers Series FED
1 options
Catalog No. 08-111-491 Supplier BINDER™ Supplier No. 80121264
Add to Cart
Add to Cart
For Use With: FED 56, FED 115, FED 260, FED 400, FED 720
  • For use on top
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
Description Access port with silicone plug - 10 mm, top
For Use With (Equipment) BINDER Chambers Series FED
Material Steel
Type Access Port
Includes Silicone plug
Diameter (Metric) Inside 10 mm