Learn More
Chemglass Life Sciences Top Adapter, #30 O-Ring Joint - For Pressure Filter Funnels (Chemglass CG-1405-20)
SDP
Catalog No. 502733594
Encompass_Preferred
The Chemglass Top Adapter (CG-1405-20) is a replacement top adapter for pressure filter funnels that uses an o-ring joint to create a larger, grease-free connection between the funnel and adapter. It is intended for use in positive-pressure filtration assemblies and pairs with compatible o-rings and pinch clamps to form a sealed, reusable connection.
  • Provides a grease-free o-ring joint connection between funnel and adapter.
  • Compatible with 30 o-ring joint sizes and standard fluoroelastomer o-rings.
  • Designed for use in positive-pressure filtration assemblies.
  • Works with stainless steel pinch clamps for secure attachment.
  • Supplies only the top adapter; o-ring and clamp sold separately.
Catalog No. 50-273-3594 Supplier Chemglass Life Sciences Supplier No. CG140520
The Chemglass Top Adapter (CG-1405-20) is a replacement top adapter for pressure filter funnels that uses an o-ring joint to create a larger, grease-free connection between the funnel and adapter. It is intended for use in positive-pressure filtration assemblies and pairs with compatible o-rings and pinch clamps to form a sealed, reusable connection.
  • Provides a grease-free o-ring joint connection between funnel and adapter.
  • Compatible with 30 o-ring joint sizes and standard fluoroelastomer o-rings.
  • Designed for use in positive-pressure filtration assemblies.
  • Works with stainless steel pinch clamps for secure attachment.
  • Supplies only the top adapter; o-ring and clamp sold separately.