Learn More
Chemglass Life Sciences Top Adapter Only, #50 O-ring Jt
SDP
Catalog No. CG140522
Encompass_Preferred
Top Adapter Only, #50 O-ring Jt. For positive pressure filtration of materials up to 15 psi. Connection between the funnel and adapter is made by an o-ring joint which permits a larger, grease free opening for sample retrieval. The 24/40 lower inner standard taper joint has an outlet for connection to a vacuum source.
Catalog No. CG140522 Supplier Chemglass Life Sciences Supplier No. CG140522
Top Adapter Only, #50 O-ring Jt. For positive pressure filtration of materials up to 15 psi. Connection between the funnel and adapter is made by an o-ring joint which permits a larger, grease free opening for sample retrieval. The 24/40 lower inner standard taper joint has an outlet for connection to a vacuum source.