Learn More
Synthware™ Top Adapter for Vacuum Trap Body
GREENER_CHOICESDP
Catalog No. 31501799 Shop All Kemtech Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
31-501-799 1 Ea.
1 options
Catalog No. 31-501-799 Supplier Synthware™ Supplier No. V175037
Add to Cart
Add to Cart
Spherical joints add flexibility to the Vacuum Trap.
Specifications
For Use With (Application) Organic Synthesis
For Use With (Equipment) Vacuum Trap
Material Borosilicate Glass
Product Type Top Adapter
Quantity 1 Ea.
Standard Taper Joint No. 35/20
Width (Metric) 12 mm